Product Details

SNP ID
rs10109778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38913890 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTTTCTGAACTCAAACTTGCTCC[A/G]TTGTGAAACTGGAGCTGGGGCGGAA
Phenotype
MIM: 607773
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLEKHA2 PubMed Links

Gene Details

Gene
PLEKHA2
Gene Name
pleckstrin homology domain containing A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021623.1 Intron NP_067636.1
XM_011544605.2 Intron XP_011542907.1
XM_011544606.2 Intron XP_011542908.1
XM_011544607.2 Intron XP_011542909.1
XM_017013718.1 Intron XP_016869207.1
XM_017013719.1 Intron XP_016869208.1
XM_017013720.1 Intron XP_016869209.1

View Full Product Details