Product Details

SNP ID
rs2283617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18595694 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGCCCAAGGCAAGTGGGATTCTC[A/G]ACCCCACCTTCCAGATGGGGAAACG
Phenotype
MIM: 604237
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf60 PubMed Links

Gene Details

Gene
C19orf60
Gene Name
chromosome 19 open reading frame 60
There are no transcripts associated with this gene.

Gene
CRLF1
Gene Name
cytokine receptor like factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004750.4 Intron NP_004741.1
XM_011528422.2 Intron XP_011526724.1
XM_011528423.2 Intron XP_011526725.1
XM_011528424.2 Intron XP_011526726.1

View Full Product Details