Product Details

SNP ID
rs111033552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116120105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCACTAGGAATCCTGAGAAAG[A/G]GGAGTGGACATACTCAGAGGAGTAT
Phenotype
MIM: 120110
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL10A1 PubMed Links

Gene Details

Gene
COL10A1
Gene Name
collagen type X alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000493.3 5554 Missense Mutation CCT,TCT P671S NP_000484.2
XM_006715333.3 5554 Missense Mutation CCT,TCT P671S XP_006715396.1
XM_011535432.2 5554 Missense Mutation CCT,TCT P671S XP_011533734.1
XM_011535433.2 5554 Missense Mutation CCT,TCT P671S XP_011533735.1
XM_017010248.1 5554 Missense Mutation CCT,TCT P671S XP_016865737.1
XM_017010249.1 5554 Missense Mutation CCT,TCT P671S XP_016865738.1
Gene
NT5DC1
Gene Name
5'-nucleotidase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152729.2 5554 Intron NP_689942.2
XM_006715378.3 5554 Intron XP_006715441.1

View Full Product Details