Product Details
- SNP ID
-
rs555551
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:119045594 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATGGGGAAAACAGAGGTGGGGGT[A/T]ATAGTTCGTCGTTCTTCAAAGGCCG
- Phenotype
-
MIM: 601746
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HYOU1
PubMed Links
Gene Details
- Gene
- HYOU1
- Gene Name
- hypoxia up-regulated 1
There are no transcripts associated with this gene.
View Full Product Details