Product Details
- SNP ID
-
rs681099
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:108206663 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACACACACATACACACGCACGCACG[C/T]ACAGTGGCCTGCACCTATAGTCCCA
- Phenotype
-
MIM: 602210
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EIF3E
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140964651] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EIF3E
- Gene Name
- eukaryotic translation initiation factor 3 subunit E
View Full Product Details