Product Details

SNP ID
rs221070
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69470390 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCTCCCCCGGAAGTGCTTTTCCA[A/G]GATTCGGGCCGGAGAGAGGCCTTGT
Phenotype
MIM: 607743
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FRS2 PubMed Links

Gene Details

Gene
FRS2
Gene Name
fibroblast growth factor receptor substrate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042555.2 42 UTR 5 NP_001036020.1
NM_001278351.1 42 UTR 5 NP_001265280.1
NM_001278353.1 42 UTR 5 NP_001265282.1
NM_001278354.1 42 UTR 5 NP_001265283.1
NM_001278355.1 42 UTR 5 NP_001265284.1
NM_001278356.1 42 UTR 5 NP_001265285.1
NM_001278357.1 42 UTR 5 NP_001265286.1
NM_006654.4 42 UTR 5 NP_006645.3
XM_017018717.1 42 UTR 5 XP_016874206.1
XM_017018718.1 42 UTR 5 XP_016874207.1
XM_017018719.1 42 Intron XP_016874208.1

View Full Product Details