Product Details

SNP ID
rs510261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60940373 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAACTGCTTTAAGGATATAGCTGG[A/G]ATAAAATCTAGTTGGGTTTTCACAC
Phenotype
MIM: 610408
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC15A3 PubMed Links

Gene Details

Gene
SLC15A3
Gene Name
solute carrier family 15 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016582.2 Intron NP_057666.1
XM_011545095.2 Intron XP_011543397.1
Gene
TMEM132A
Gene Name
transmembrane protein 132A
There are no transcripts associated with this gene.

View Full Product Details