Product Details

SNP ID
rs319597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134905755 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCTGCTGTAGAAATTAATATTTT[A/G]GAGTAAATGGAGGGAAAGTTTTAAC
Phenotype
MIM: 609836
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCBD2 PubMed Links

Gene Details

Gene
PCBD2
Gene Name
pterin-4 alpha-carbinolamine dehydratase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032151.4 Intron NP_115527.3
Gene
TXNDC15
Gene Name
thioredoxin domain containing 15
There are no transcripts associated with this gene.

View Full Product Details