Product Details

SNP ID
rs552560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125497577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAATAAATAAATAAATAAATCAC[A/C]AAGAGTAGCTGGTTAAAAGATCCCT
Phenotype
MIM: 604825
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FEZ1 PubMed Links

Gene Details

Gene
FEZ1
Gene Name
fasciculation and elongation protein zeta 1
There are no transcripts associated with this gene.

Gene
LOC403312
Gene Name
putative uncharacterized protein MGC39545
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301851.1 1472 UTR 3 NP_001288780.1

View Full Product Details