Product Details
- SNP ID
-
rs336362
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:142023948 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGGATAGCTAAATTCATTACATTT[C/T]TTTTTTTGCTTATATTACCTTACTA
- Phenotype
-
MIM: 607494
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
INPP4B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs543863678] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- INPP4B
- Gene Name
- inositol polyphosphate-4-phosphatase type II B
View Full Product Details