Product Details

SNP ID
rs413390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:371723 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCGCACAGGCACCTGTGGTCTCC[A/C]AGTAACCAAGCCCAGGCCCCAGGGG
Phenotype
MIM: 604853
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MRPL28 PubMed Links

Gene Details

Gene
MRPL28
Gene Name
mitochondrial ribosomal protein L28
There are no transcripts associated with this gene.

Gene
TMEM8A
Gene Name
transmembrane protein 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021259.2 2709 UTR 3 NP_067082.2

View Full Product Details