Product Details

SNP ID
rs625531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:74538785 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTCTGCCTGATGTCAAGGGCTTC[C/T]TGGACAGTTTGGACGTTACAGTTCG
Phenotype
MIM: 615579
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATXN7L3B PubMed Links

Gene Details

Gene
ATXN7L3B
Gene Name
ataxin 7 like 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136262.1 1015 UTR 3 NP_001129734.1

View Full Product Details