Product Details

SNP ID
rs285269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16200686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACACCAGCGTGGCCACACTGG[C/G]CTGGGAGGAAGCTGTCCTAGTAGGA
Phenotype
MIM: 603535 MIM: 614554
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AP1M1 PubMed Links

Gene Details

Gene
AP1M1
Gene Name
adaptor related protein complex 1 mu 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130524.1 Intron NP_001123996.1
NM_032493.3 Intron NP_115882.1
XM_017027420.1 Intron XP_016882909.1
XM_017027421.1 Intron XP_016882910.1
Gene
FAM32A
Gene Name
family with sequence similarity 32 member A
There are no transcripts associated with this gene.

View Full Product Details