Product Details

SNP ID
rs671386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:10670602 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGAAGGAAAATGTCACATACTGG[A/G]AAAACCATTGCCTTAAATCATGGAA
Phenotype
MIM: 613629
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927410 PubMed Links

Gene Details

Gene
LOC101927410
Gene Name
uncharacterized LOC101927410
There are no transcripts associated with this gene.

Gene
PIEZO2
Gene Name
piezo type mechanosensitive ion channel component 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022068.3 10288 UTR 3 NP_071351.2
XM_011525723.2 10288 UTR 3 XP_011524025.1
XM_011525724.2 10288 UTR 3 XP_011524026.1
XM_011525725.2 10288 UTR 3 XP_011524027.1
XM_011525726.2 10288 UTR 3 XP_011524028.1
XM_017025918.1 10288 UTR 3 XP_016881407.1

View Full Product Details