Product Details

SNP ID
rs624856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:93747542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCTATCTATATCTTACTACTTA[C/T]GTATATCTATATCTTACTTATCAAA
Phenotype
MIM: 615810 MIM: 612823
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf54 PubMed Links

Gene Details

Gene
C11orf54
Gene Name
chromosome 11 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286067.1 Intron NP_001272996.1
NM_001286068.1 Intron NP_001272997.1
NM_001286069.1 Intron NP_001272998.1
NM_001286070.1 Intron NP_001272999.1
NM_001286071.1 Intron NP_001273000.1
NM_014039.3 Intron NP_054758.2
XM_006718824.1 Intron XP_006718887.1
XM_011542781.2 Intron XP_011541083.1
XM_011542782.2 Intron XP_011541084.1
XM_017017613.1 Intron XP_016873102.1
XM_017017614.1 Intron XP_016873103.1
XM_017017615.1 Intron XP_016873104.1
XM_017017616.1 Intron XP_016873105.1
XM_017017617.1 Intron XP_016873106.1
XM_017017618.1 Intron XP_016873107.1
XM_017017619.1 Intron XP_016873108.1
Gene
TAF1D
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit D
There are no transcripts associated with this gene.

View Full Product Details