Product Details

SNP ID
rs526814
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:177176821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCCAAAGCCTTTGGGAAGCATT[A/G]CAGCTCTCAGCTCACATACAAAGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRINP2 PubMed Links

Gene Details

Gene
BRINP2
Gene Name
BMP/retinoic acid inducible neural specific 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021165.3 Intron NP_066988.1
XM_005245379.1 Intron XP_005245436.1
XM_011509826.1 Intron XP_011508128.1

View Full Product Details