Product Details

SNP ID
rs11821492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130678403 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTTGCATGTGGTGGTTTCATTAC[C/T]TCTATCAGCTGAGTCTACTGTGTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf44 PubMed Links

Gene Details

Gene
C11orf44
Gene Name
chromosome 11 open reading frame 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271983.1 Intron NP_001258912.1

View Full Product Details