Product Details

SNP ID
rs34161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:31688135 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGTCCCTCACACTCACGAAAAT[A/G]TCAGTGTGTAGGACCTGGGCTCGGG
Phenotype
MIM: 602987 MIM: 604088
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDE1C PubMed Links

Gene Details

Gene
PDE1C
Gene Name
phosphodiesterase 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191056.3 Intron NP_001177985.1
NM_001191057.3 Intron NP_001177986.1
NM_001191058.3 Intron NP_001177987.2
NM_001191059.3 Intron NP_001177988.1
NM_001322055.1 Intron NP_001308984.1
NM_001322056.1 Intron NP_001308985.1
NM_001322057.1 Intron NP_001308986.1
NM_001322058.1 Intron NP_001308987.1
NM_001322059.1 Intron NP_001308988.1
NM_005020.4 Intron NP_005011.1
XM_017012264.1 Intron XP_016867753.1
XM_017012265.1 Intron XP_016867754.1
XM_017012266.1 Intron XP_016867755.1
XM_017012267.1 Intron XP_016867756.1
Gene
PPP1R17
Gene Name
protein phosphatase 1 regulatory subunit 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145123.2 Intron NP_001138595.1
NM_006658.4 Intron NP_006649.2
XM_011515094.1 Intron XP_011513396.1

View Full Product Details