Gene Transcripts
- Gene Symbol
- TMEM50B
- Entrez Gene ID
- 757
- Gene Name
- transmembrane protein 50B
- Gene Aliases
- C21orf4, HCVP7TP3
- Chromosome Location
- Chr. 21: 33432486 - 33480009 on Build GRCh38
- UniGene ID
- Hs.433668
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006134.6 |
NP_006125.2
|
3, 4 |
436 |
|
NR_040016.1 |
|
3, 4 |
436 |
|
XM_011529746.1 |
XP_011528048.1
|
3, 4 |
434 |
|
GenBank mRNA |
AF045606.2 |
- |
3, 4 |
381 |
|
AF086280.1 |
- |
2, 3 |
270 |
|
AK309977.1 |
- |
3, 4 |
363 |
|
AK310701.1 |
- |
3, 4 |
292 |
|
AK311870.1 |
- |
2, 3 |
256 |
|
AY358634.1 |
- |
3, 4 |
360 |
|
AY820138.1 |
- |
2, 3 |
223 |
|
BC000569.2 |
- |
3, 4 |
352 |
3342648 |
DA286739.1 |
- |
3, 4 |
437 |
|
DQ286229.1 |
- |
2, 3 |
223 |
|
View Full Product Details