Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_011527533.2 | XP_011525835.1 | 5 | 1231 | |
XM_011527534.2 | XP_011525836.1 | 5 | 1234 | ||
XM_011527535.2 | XP_011525837.1 | 5 | 1145 | ||
XM_011527536.2 | XP_011525838.1 | 5 | 1070 | ||
XM_011527537.2 | XP_011525839.1 | 5 | 1037 | ||
XM_011527538.2 | XP_011525840.1 | 5 | 1233 | ||
XM_017027511.1 | XP_016883000.1 | 5 | 1234 | ||
GenBank mRNA | AK300170.1 | - | 4 | 990 | |
AK300182.1 | - | 5 | 1169 | ||
AK308174.1 | - | 5 | 1145 | ||
AK309903.1 | - | 5 | 1037 | ||
BC035359.1 | - | 4 | 1011 | 5176194 | |
BC140798.1 | - | 5 | 976 | 9021666 | |
CA495365.1 | - | 3 | 696 | ||
D86358.1 | - | 5 | 1193 | ||
D86359.1 | - | 5 | 1193 | ||
U71382.1 | - | 5 | 987 |