Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001962.2 | NP_001953.1 | 2 | 499 | |
XM_006714565.2 | XP_006714628.1 | 2 | 1968 | ||
XM_011543250.2 | XP_011541552.1 | 2 | 312 | ||
XM_011543251.2 | XP_011541553.1 | 2 | 1825 | ||
XM_017009205.1 | XP_016864694.1 | 2 | 1588 | ||
GenBank mRNA | AK308672.1 | - | 2 | 489 | |
BC075054.2 | - | 2 | 266 | 30915365 | |
BC075055.2 | - | 2 | 266 | 30915581 | |
U26403.1 | - | 2 | 499 |