Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006151.2 | NP_006142.1 | 4 | 588 | |
NR_027647.1 | 3 | 500 | |||
XM_011524808.2 | XP_011523110.1 | 4 | 464 | ||
GenBank mRNA | AK131496.1 | - | 3 | 1251 | |
AK303903.1 | - | 3 | 335 | ||
AK308378.1 | - | 4 | 313 | ||
AK310965.1 | - | 4 | 343 | ||
BC107166.1 | - | 4 | 459 | 40034365 | |
BC107167.1 | - | 4 | 459 | 40034371 | |
BX648309.1 | - | 4 | 426 | ||
U39573.1 | - | 4 | 353 |