Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002775.4 | NP_002766.1 | 9 | 1471 | |
GenBank mRNA | AF070555.1 | - | 7 | 745 | |
AF097709.1 | - | 9 | 914 | ||
AK091944.1 | - | 11 | 1246 | ||
AK092476.1 | - | 10 | 1252 | ||
AK290089.1 | - | 9 | 962 | ||
BC011352.1 | - | 10 | 1238 | ||
BC031082.1 | - | 9 | 1379 | ||
D87258.1 | - | 9 | 1391 | ||
Y07921.1 | - | 9 | 1381 |