Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002889.3 | NP_002880.1 | 5 | 564 | |
GenBank mRNA | AB015632.1 | - | 5 | 519 | |
AK092804.1 | - | 3 | 1731 | ||
AK312197.1 | - | 5 | 518 | ||
BC000069.2 | - | 5 | 495 | 3506067 | |
BM919890.1 | - | 4 | 499 | ||
BX448753.2 | - | 5 | 563 | ||
CR541992.1 | - | 4 | 447 | ||
CR542026.1 | - | 4 | 447 | ||
U77594.2 | - | 5 | 507 |