Gene Transcripts

Gene Symbol
STK11
Entrez Gene ID
6794
Gene Name
serine/threonine kinase 11
Gene Aliases
LKB1, PJS, hLKB1
Chromosome Location
Chr. 19: 1205799 - 1228435 on Build GRCh38
UniGene ID
Hs.515005
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000455.4 NP_000446.1 1, 2 1416
XM_005259617.2 XP_005259674.1 1, 2 926
XM_005259618.3 XP_005259675.1 1, 2 926
XM_011528209.1 XP_011526511.1 2, 3 1057
GenBank mRNA AB209553.1 - 1, 2 391
AF035625.1 - 1, 2 639
AF217978.1 - 2, 3 1262
AK314858.1 - 1, 2 427
BC007981.2 - 1, 2 751 3689780
BC019334.1 - 1, 2 572 3946005
EF644172.1 - 1, 2 385
KU178339.1 - 1, 2 301
U63333.1 - 1, 2 301

View Full Product Details