Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142391.2 | NP_001135863.1 | 4 | 837 | |
NM_001142392.2 | NP_001135864.1 | 3 | 924 | ||
NM_019848.4 | NP_062822.1 | 2 | 1159 | ||
XM_005277913.4 | XP_005277970.1 | 2 | 1581 | ||
XM_006724847.3 | XP_006724910.1 | 2 | 1102 | ||
XM_006724848.3 | XP_006724911.1 | 3 | 1015 | ||
XM_011531200.2 | XP_011529502.2 | 2 | 1134 | ||
XM_011531201.2 | XP_011529503.1 | 2 | 838 | ||
GenBank mRNA | BC004966.1 | - | 4 | 761 | |
BX401493.2 | - | 3 | 896 |