Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004855.4 | NP_004846.4 | 9 | 1391 | |
XM_005254795.4 | XP_005254852.1 | 7 | 820 | ||
XM_011522235.2 | XP_011520537.1 | 9 | 1346 | ||
XM_011522236.2 | XP_011520538.1 | 8 | 1286 | ||
XM_011522237.2 | XP_011520539.1 | 8 | 895 | ||
XM_017022730.1 | XP_016878219.1 | 8 | 810 | ||
XM_017022731.1 | XP_016878220.1 | 9 | 939 | ||
XM_017022732.1 | XP_016878221.1 | 7 | 775 | ||
GenBank mRNA | AK223330.1 | - | 9 | 1096 | |
AK293666.1 | - | 6 | 1007 | ||
AK314706.1 | - | 9 | 1366 | ||
BC017711.1 | - | 9 | 1328 | 4430974 | |
D42138.1 | - | 9 | 1119 |