Gene Transcripts

Gene Symbol
BRD8
Entrez Gene ID
10902
Gene Name
bromodomain containing 8
Gene Aliases
SMAP, SMAP2, p120
Chromosome Location
Chr. 5: 138139766 - 138178669 on Build GRCh38
UniGene ID
Hs.519337
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164326.1 NP_001157798.1 12 1666
NM_001300961.1 NP_001287890.1 12 1662
NM_001300962.1 NP_001287891.1 11 1587
NM_001300966.1 NP_001287895.1 13 1754
NM_006696.3 NP_006687.3 13 1876
NM_139199.1 NP_631938.1 12 1657
XM_005271855.2 XP_005271912.1 13 1867
XM_005271856.2 XP_005271913.1 13 1867
XM_005271857.2 XP_005271914.1 12 1657
XM_005271859.3 XP_005271916.1 11 1438
XM_005271860.3 XP_005271917.1 13 1867
XM_005271861.3 XP_005271918.1 12 1648
XM_011543109.1 XP_011541411.1 13 1867
XM_011543110.1 XP_011541412.1 13 1853
XM_017008969.1 XP_016864458.1 12 1648
XM_017008970.1 XP_016864459.1 13 1867
XM_017008971.1 XP_016864460.1 13 1867
XM_017008972.1 XP_016864461.1 12 1657
XM_017008973.1 XP_016864462.1 12 1648
XM_017008974.1 XP_016864463.1 12 1657
XM_017008975.1 XP_016864464.1 12 1648
XM_017008976.1 XP_016864465.1 11 1438
GenBank mRNA AB209079.1 - 13 1822
AF016270.1 - 12 1808
AK225928.1 - 12 1629
AK289951.1 - 13 1836
AK293621.1 - 11 1547
AK297608.1 - 3 945
AK297756.1 - 12 1622
AK312333.1 - 13 1836
AL136823.1 - 12 1630
BC008039.2 - 13 1818 3009915
BC008076.2 - 13 1818 3009915
X87613.1 - 12 1684
X94234.1 - 11 1596

View Full Product Details