Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020650.2 | NP_065701.2 | 7 | 1348 | |
XM_005259089.3 | XP_005259146.1 | 8 | 1443 | ||
XM_011527143.1 | XP_011525445.1 | 8 | 1537 | ||
XM_017027023.1 | XP_016882512.1 | 7 | 1622 | ||
GenBank mRNA | AF183423.1 | - | 6 | 933 | |
AY195859.1 | - | 7 | 1348 | ||
AY358423.1 | - | 7 | 935 | ||
BC013436.1 | - | 7 | 931 | 3889718 |