Gene Transcripts
- Gene Symbol
- RXFP1
- Entrez Gene ID
- 59350
- Gene Name
- relaxin/insulin like family peptide receptor 1
- Gene Aliases
- LGR7, RXFPR1
- Chromosome Location
- Chr. 4: 158521711 - 158653372 on Build GRCh38
- UniGene ID
- Hs.591686
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253727.1 |
NP_001240656.1
|
4, 5 |
742 |
|
NM_001253728.1 |
NP_001240657.1
|
3, 4 |
562 |
|
NM_001253729.1 |
NP_001240658.1
|
4, 5 |
661 |
|
NM_001253730.1 |
NP_001240659.1
|
3, 4 |
586 |
|
NM_001253732.1 |
NP_001240661.1
|
4, 5 |
661 |
|
NM_001253733.1 |
NP_001240662.1
|
4, 5 |
661 |
|
NM_021634.3 |
NP_067647.2
|
4, 5 |
661 |
|
NR_045579.1 |
|
5, 6 |
1350 |
|
NR_045580.1 |
|
4, 5 |
714 |
|
NR_045581.1 |
|
4, 5 |
685 |
|
NR_045582.1 |
|
4, 5 |
766 |
|
NR_045583.1 |
|
3, 4 |
604 |
|
NR_045584.1 |
|
4, 5 |
661 |
|
XM_011532174.1 |
XP_011530476.1
|
4, 5 |
711 |
|
XM_011532175.1 |
XP_011530477.1
|
4, 5 |
711 |
|
XM_011532176.1 |
XP_011530478.1
|
4, 5 |
633 |
|
XM_011532179.2 |
XP_011530481.1
|
4, 5 |
711 |
|
XM_017008517.1 |
XP_016864006.1
|
4, 5 |
711 |
|
XM_017008518.1 |
XP_016864007.1
|
4, 5 |
633 |
|
XM_017008519.1 |
XP_016864008.1
|
5, 6 |
675 |
|
XM_017008520.1 |
XP_016864009.1
|
4, 5 |
764 |
|
XM_017008521.1 |
XP_016864010.1
|
5, 6 |
594 |
|
XM_017008522.1 |
XP_016864011.1
|
4, 5 |
682 |
|
XM_017008523.1 |
XP_016864012.1
|
4, 5 |
633 |
|
XM_017008524.1 |
XP_016864013.1
|
4, 5 |
633 |
|
XM_017008525.1 |
XP_016864014.1
|
3, 4 |
565 |
|
GenBank mRNA |
AB208943.1 |
- |
4, 5 |
399 |
|
AF190500.1 |
- |
4, 5 |
398 |
|
AK122647.1 |
- |
6, 7 |
702 |
|
AK294694.1 |
- |
4, 5 |
506 |
|
AK295040.1 |
- |
5, 6 |
1173 |
|
AK300379.1 |
- |
3, 4 |
457 |
|
AK307889.1 |
- |
3, 4 |
503 |
|
AK312470.1 |
- |
4, 5 |
493 |
|
AY899848.1 |
- |
4, 5 |
398 |
|
AY899849.1 |
- |
4, 5 |
398 |
|
AY899850.1 |
- |
3, 4 |
299 |
|
BC112142.1 |
- |
4, 5 |
480 |
8327610 |
BC113617.1 |
- |
4, 5 |
483 |
8322669 |
BC143556.1 |
- |
4, 5 |
503 |
|
BC143558.1 |
- |
4, 5 |
536 |
|
BC143559.1 |
- |
3, 4 |
408 |
|
BX647975.1 |
- |
4, 5 |
599 |
|
BX647980.1 |
- |
4, 5 |
599 |
|
BX647985.1 |
- |
4, 5 |
628 |
|
CR748189.1 |
- |
1, 2 |
33 |
|
View Full Product Details