Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172173.1 | NP_001165644.1 | 7 | 2002 | |
NM_024969.3 | NP_079245.2 | 5 | 1901 | ||
XM_005246865.4 | XP_005246922.1 | 5 | 1724 | ||
GenBank mRNA | AB063300.1 | - | 5 | 1962 | |
AK021765.1 | - | 2 | 1035 | ||
AK056655.1 | - | 5 | 1898 | ||
AK126129.1 | - | 8 | 1791 |