Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031907.1 | NP_114113.1 | 1 | 132 | |
XM_017029892.1 | XP_016885381.1 | 2 | 218 | ||
GenBank mRNA | AF285593.1 | - | 1 | 132 | |
AK292527.1 | - | 6 | 550 | ||
BC069073.1 | - | 1 | 131 | 7216887 | |
BC101189.2 | - | 1 | 130 | 40019498 | |
BC101190.2 | - | 1 | 130 | 40019499 | |
BC101191.2 | - | 1 | 130 | 40019500 |