Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004918.3 | NP_004909.1 | 1 | 64 | |
GenBank mRNA | AB018563.1 | - | 1 | 43 | |
AF110466.1 | - | 1 | 40 | ||
AF137027.1 | - | 1 | 59 | ||
BC041616.1 | - | 1 | 38 | 5816694 | |
BC051000.1 | - | 1 | 64 | 5813497 | |
CR457087.1 | - | 1 | 13 |