Gene Transcripts

Gene Symbol
OR10H2
Entrez Gene ID
26538
Gene Name
olfactory receptor family 10 subfamily H member 2
Gene Aliases
Chromosome Location
Chr. 19: 15728024 - 15729052 on Build GRCh38
UniGene ID
Hs.247694
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013939.2 NP_039227.1 1 41
GenBank mRNA BC069085.1 - 1 21
BC069457.1 - 1 21 7262254
BC112178.1 - 1 53 8327646
BC113647.1 - 1 41 8322699

View Full Product Details