Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001173516.1 | NP_001166987.1 | 7, 8 | 849 | |
NM_001173517.1 | NP_001166988.1 | 6, 7 | 856 | ||
NM_024597.3 | NP_078873.2 | 7, 8 | 961 | ||
XM_005262472.1 | XP_005262529.1 | 7, 8 | 816 | ||
XM_017029843.1 | XP_016885332.1 | 7, 8 | 813 | ||
GenBank mRNA | AK301478.1 | - | 7, 8 | 850 | |
AL832120.1 | - | 6, 7 | 648 | ||
BC040518.1 | - | 7, 8 | 825 |