Gene Transcripts

Gene Symbol
YDJC
Entrez Gene ID
150223
Gene Name
YdjC homolog (bacterial)
Gene Aliases
Chromosome Location
Chr. 22: 21628089 - 21630326 on Build GRCh38
UniGene ID
Hs.355952
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017964.1 NP_001017964.1 2 306
XM_011529906.2 XP_011528208.1 2 581
GenBank mRNA BC096754.1 - 2 320
BC110075.1 - 2 290
BC110799.1 - 2 291
BC133043.1 - 2 401
BC133047.1 - 2 401
BI597711.1 - 2 325

View Full Product Details