Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005270575.3 | XP_005270632.1 | 20 | 23323 | |
XM_005270576.3 | XP_005270633.1 | 14 | 18107 | ||
XM_011540879.2 | XP_011539181.1 | 19 | 18751 | ||
XM_011540882.2 | XP_011539184.1 | 16 | 16966 | ||
XM_017000523.1 | XP_016856012.1 | 20 | 23339 | ||
XM_017000524.1 | XP_016856013.1 | 15 | 17137 | ||
XM_017000525.1 | XP_016856014.1 | 16 | 16971 | ||
XM_017000526.1 | XP_016856015.1 | 15 | 16930 | ||
XM_017000527.1 | XP_016856016.1 | 15 | 17109 | ||
XM_017000528.1 | XP_016856017.1 | 16 | 17058 |