Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_024271.1 | 1 | 305 | ||
GenBank mRNA | AL832059.1 | - | 1 | 241 | |
BC092503.1 | - | 1 | 250 | ||
DA100871.1 | - | 1 | 305 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_136590.1 | 1 | 264 | ||
NR_136591.1 | 1 | 264 | |||
NR_136592.1 | 1 | 264 | |||
NR_136593.1 | 1 | 264 | |||
GenBank mRNA | DA132394.1 | - | 1 | 247 | |
DB083170.1 | - | 1 | 265 |