Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007197.3 | NP_009128.1 | 1 | 1989 | |
GenBank mRNA | AB027464.1 | - | 1 | 1522 | |
BC070037.1 | - | 1 | 1990 | 30337870 | |
BC074997.2 | - | 1 | 1526 | 30915397 | |
BC074998.2 | - | 1 | 1526 | 30915609 |