Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130003.1 | NP_001123475.1 | 3 | 442 | |
NM_144642.4 | NP_653243.1 | 2 | 455 | ||
XM_017005731.1 | XP_016861220.1 | 3 | 208 | ||
XM_017005732.1 | XP_016861221.1 | 3 | 442 | ||
GenBank mRNA | AF411860.2 | - | 2 | 204 | |
AK122831.1 | - | 3 | 232 | ||
AK293628.1 | - | 3 | 353 | ||
AK312465.1 | - | 2 | 360 | ||
AK315958.1 | - | 2 | 418 | ||
AL834457.1 | - | 2 | 184 | ||
BC022518.1 | - | 2 | 204 | 4792167 | |
CR936762.1 | - | 3 | 363 | ||
DA505318.1 | - | 2 | 456 |