Gene Transcripts

Gene Symbol
CYLC2
Entrez Gene ID
1539
Gene Name
cylicin 2
Gene Aliases
Chromosome Location
Chr. 9: 102995311 - 103018488 on Build GRCh38
UniGene ID
Hs.3232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001340.4 NP_001331.1 4 328
GenBank mRNA AK313349.1 - 4 315
BC058038.1 - 4 332
BC114547.1 - 4 261 40005379
Z46788.1 - 4 261

View Full Product Details