Gene Transcripts
- Gene Symbol
- UHRF1BP1L
- Entrez Gene ID
- 23074
- Gene Name
- UHRF1 binding protein 1 like
- Gene Aliases
- SHIP164
- Chromosome Location
- Chr. 12: 100026251 - 100142864 on Build GRCh38
- UniGene ID
- Hs.572154
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001006947.1 |
NP_001006948.1
|
9, 10 |
1342 |
|
NM_015054.1 |
NP_055869.1
|
9, 10 |
1342 |
|
XM_005268738.2 |
XP_005268795.1
|
10, 11 |
1448 |
|
XM_017019047.1 |
XP_016874536.1
|
9, 10 |
1302 |
|
XM_017019049.1 |
XP_016874538.1
|
9, 10 |
1302 |
|
XM_017019050.1 |
XP_016874539.1
|
9, 10 |
1302 |
|
XM_017019051.1 |
XP_016874540.1
|
10, 11 |
1448 |
|
GenBank mRNA |
AB014601.2 |
- |
5, 6 |
783 |
|
AK125813.1 |
- |
9, 10 |
1302 |
|
AL136821.1 |
- |
2, 3 |
147 |
|
BC014891.1 |
- |
9, 10 |
1325 |
|
BC026286.1 |
- |
2, 3 |
109 |
|
BC127017.1 |
- |
9, 10 |
1160 |
40124660 |
BX647380.1 |
- |
3, 4 |
313 |
|
View Full Product Details