Gene Transcripts

Gene Symbol
SHC2
Entrez Gene ID
25759
Gene Name
SHC adaptor protein 2
Gene Aliases
SCK, SHCB, SLI
Chromosome Location
Chr. 19: 416583 - 461011 on Build GRCh38
UniGene ID
Hs.30965
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012435.2 NP_036567.2 5, 6 774
XM_011527893.2 XP_011526195.1 5, 6 789
XM_011527894.2 XP_011526196.1 5, 6 789
XM_011527895.2 XP_011526197.1 5, 6 789
XM_011527896.2 XP_011526198.1 7, 8 834
XM_017026567.1 XP_016882056.1 6, 7 922
GenBank mRNA AB001451.1 - 5, 6 648
AL134433.1 - 5, 6 431
AL360254.1 - 3, 4 204

View Full Product Details