Gene Transcripts

Gene Symbol
HINFP
Entrez Gene ID
25988
Gene Name
histone H4 transcription factor
Gene Aliases
HiNF-P, MIZF, ZNF743
Chromosome Location
Chr. 11: 119121523 - 119135055 on Build GRCh38
UniGene ID
Hs.504091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243259.1 NP_001230188.1 4 609
NM_015517.4 NP_056332.2 5 726
NM_198971.2 NP_945322.1 4 609
XM_011542744.2 XP_011541046.1 4 718
XM_011542745.2 XP_011541047.1 4 568
XM_011542746.1 XP_011541048.1 3 389
XM_017017499.1 XP_016872988.1 4 666
XM_017017500.1 XP_016872989.1 4 545
XM_017017501.1 XP_016872990.1 5 672
XM_017017502.1 XP_016872991.1 7 997
XM_017017503.1 XP_016872992.1 5 762
XM_017017504.1 XP_016872993.1 5 637
XM_017017505.1 XP_016872994.1 4 560
XM_017017506.1 XP_016872995.1 4 523
XM_017017507.1 XP_016872996.1 6 762
XM_017017508.1 XP_016872997.1 5 637
XM_017017509.1 XP_016872998.1 3 389
GenBank mRNA AK056362.1 - 4 522
AK092869.1 - 1 167
AK301452.1 - 4 516
BC001073.2 - 4 580 2820431
BC001945.1 - 4 580 2820431
BC012856.1 - 4 521 3871231
BC017234.2 - 4 529 3537381

View Full Product Details