Gene Transcripts

Gene Symbol
LAMA5
Entrez Gene ID
3911
Gene Name
laminin subunit alpha 5
Gene Aliases
Chromosome Location
Chr. 20: 62309060 - 62367318 on Build GRCh38
UniGene ID
Hs.473256
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005560.4 NP_005551.3 35 4646
XM_006723796.3 XP_006723859.1 35 4649
XM_006723798.3 XP_006723861.1 35 4652
XM_011528818.2 XP_011527120.1 34 4511
XM_011528819.2 XP_011527121.1 33 4376
GenBank mRNA AB010099.1 - 1 66
AB067494.1 - 32 3990
AF443072.1 - 35 4646

View Full Product Details