Gene Transcripts

Gene Symbol
UGT1A3
Entrez Gene ID
54659
Gene Name
UDP glucuronosyltransferase family 1 member A3
Gene Aliases
UDPGT, UDPGT 1-3, UGT-1C, UGT1-03, UGT1.3, UGT1A3S, UGT1C
Chromosome Location
Chr. 2: 233729127 - 233773299 on Build GRCh38
UniGene ID
Hs.554822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019093.2 NP_061966.1 1 344
GenBank mRNA AY435138.1 - 1 344

View Full Product Details