Gene Transcripts

Gene Symbol
RMDN3
Entrez Gene ID
55177
Gene Name
regulator of microtubule dynamics 3
Gene Aliases
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51
Chromosome Location
Chr. 15: 40735884 - 40755336 on Build GRCh38
UniGene ID
Hs.511067
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304802.1 NP_001291731.1 6 1123
NM_001323894.1 NP_001310823.1 6 1037
NM_001323895.1 NP_001310824.1 6 1254
NM_001323896.1 NP_001310825.1 6 1115
NM_001323897.1 NP_001310826.1 6 1231
NM_018145.2 NP_060615.1 6 1153
XM_011521755.2 XP_011520057.1 3 1399
GenBank mRNA AB000782.1 - 5 892
AK001441.1 - 6 1077
AK025963.1 - 3 466
AK092058.1 - 6 1036
AK097286.1 - 7 965
AK123192.1 - 7 1031
AK123282.1 - 5 1960
AY358793.1 - 5 893
BC008970.2 - 2 350
BC063844.1 - 6 1041 6527521

View Full Product Details