Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001322340.1 | NP_001309269.1 | 12 | 1467 | |
NM_001322341.1 | NP_001309270.1 | 13 | 1571 | ||
NM_001322342.1 | NP_001309271.1 | 12 | 971 | ||
NM_001322343.1 | NP_001309272.1 | 12 | 1030 | ||
NM_022779.8 | NP_073616.6 | 12 | 1467 | ||
NM_138620.1 | NP_619526.1 | 12 | 1467 | ||
NR_136309.1 | 12 | 1467 | |||
XM_005272207.4 | XP_005272264.1 | 12 | 1154 | ||
XM_006717236.3 | XP_006717299.1 | 12 | 1469 | ||
XM_011518921.2 | XP_011517223.1 | 12 | 1469 | ||
XM_011518922.2 | XP_011517224.1 | 12 | 1469 | ||
GenBank mRNA | AF335568.1 | - | 12 | 1468 | |
AF335569.1 | - | 12 | 1468 | ||
AF427339.1 | - | 12 | 1468 | ||
AK023695.1 | - | 8 | 540 | ||
AK027002.1 | - | 12 | 1051 | ||
AK027484.1 | - | 12 | 1468 | ||
AK225121.1 | - | 12 | 1022 | ||
AK225350.1 | - | 12 | 1052 | ||
AK302670.1 | - | 12 | 1204 | ||
BC012726.2 | - | 12 | 1093 |