Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72228242-72228744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
UniGene
Hs.523875
Gene Chromosome Location
Chr.11: 72223861-72239147 on Build GRCh38
Transcripts
NM_001567.3 XM_005273978.3 XM_005273979.4 XM_011544999.2 XM_011545000.2 XM_017017673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4036452 c.269G>A p.R90H Chr.11: 72228370-72228370 on Build GRCh38
6686199 c.306C>T p.Y102Y Chr.11: 72228407-72228407 on Build GRCh38
931658 c.252G>A p.S84S Chr.11: 72228353-72228353 on Build GRCh38
931659 c.272G>A p.R91H Chr.11: 72228373-72228373 on Build GRCh38
4882771 c.376C>A p.P126T Chr.11: 72228477-72228477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142516692 Chr.11: 72228373-72228373 on Build GRCh38
rs965895209 Chr.11: 72228270-72228270 on Build GRCh38
rs773393276 Chr.11: 72228456-72228456 on Build GRCh38
rs536046056 Chr.11: 72228617-72228617 on Build GRCh38
rs372885646 Chr.11: 72228414-72228414 on Build GRCh38
rs1048595584 Chr.11: 72228641-72228641 on Build GRCh38
rs760937587 Chr.11: 72228458-72228458 on Build GRCh38
rs770629033 Chr.11: 72228357-72228357 on Build GRCh38
rs776894351 Chr.11: 72228523-72228523 on Build GRCh38
rs750670978 Chr.11: 72228383-72228383 on Build GRCh38
rs756549919 Chr.11: 72228541-72228541 on Build GRCh38
rs756201014 Chr.11: 72228332-72228332 on Build GRCh38
rs920419704 Chr.11: 72228594-72228594 on Build GRCh38
rs767268219 Chr.11: 72228691-72228691 on Build GRCh38
rs753208760 Chr.11: 72228468-72228468 on Build GRCh38
rs528612645 Chr.11: 72228478-72228478 on Build GRCh38
rs774083889 Chr.11: 72228507-72228507 on Build GRCh38
rs753046122 Chr.11: 72228408-72228408 on Build GRCh38
rs762663532 Chr.11: 72228370-72228370 on Build GRCh38
rs748574176 Chr.11: 72228275-72228276 on Build GRCh38
rs139756712 Chr.11: 72228469-72228469 on Build GRCh38
rs745317721 Chr.11: 72228486-72228486 on Build GRCh38
rs1031114646 Chr.11: 72228510-72228512 on Build GRCh38
rs376366065 Chr.11: 72228696-72228696 on Build GRCh38
rs762050147 Chr.11: 72228313-72228313 on Build GRCh38
rs957763736 Chr.11: 72228352-72228352 on Build GRCh38
rs766486781 Chr.11: 72228396-72228396 on Build GRCh38
rs758746094 Chr.11: 72228412-72228412 on Build GRCh38
rs780526227 Chr.11: 72228333-72228333 on Build GRCh38
rs549593209 Chr.11: 72228418-72228418 on Build GRCh38
rs754410639 Chr.11: 72228341-72228341 on Build GRCh38
rs768403979 Chr.11: 72228447-72228447 on Build GRCh38
rs759655328 Chr.11: 72228532-72228532 on Build GRCh38
rs538502034 Chr.11: 72228548-72228548 on Build GRCh38
rs200615694 Chr.11: 72228536-72228536 on Build GRCh38
rs930093088 Chr.11: 72228618-72228618 on Build GRCh38
rs764732670 Chr.11: 72228547-72228547 on Build GRCh38
rs779479923 Chr.11: 72228706-72228706 on Build GRCh38
rs916778981 Chr.11: 72228337-72228337 on Build GRCh38
rs377672840 Chr.11: 72228399-72228399 on Build GRCh38
rs775305188 Chr.11: 72228435-72228435 on Build GRCh38
rs749097157 Chr.11: 72228443-72228443 on Build GRCh38
rs797044469 Chr.11: 72228379-72228383 on Build GRCh38
rs527957059 Chr.11: 72228432-72228432 on Build GRCh38
rs763316860 Chr.11: 72228546-72228546 on Build GRCh38
rs916336133 Chr.11: 72228525-72228525 on Build GRCh38
rs1801031 Chr.11: 72228425-72228425 on Build GRCh38
rs1048473891 Chr.11: 72228644-72228644 on Build GRCh38
rs978297595 Chr.11: 72228271-72228271 on Build GRCh38
rs768475256 Chr.11: 72228505-72228505 on Build GRCh38
rs749225658 Chr.11: 72228495-72228495 on Build GRCh38
rs751712397 Chr.11: 72228311-72228311 on Build GRCh38
rs770179572 Chr.11: 72228715-72228715 on Build GRCh38
rs183522478 Chr.11: 72228294-72228294 on Build GRCh38
rs146864944 Chr.11: 72228404-72228404 on Build GRCh38
rs754321111 Chr.11: 72228398-72228398 on Build GRCh38
rs746501732 Chr.11: 72228354-72228354 on Build GRCh38
rs200094851 Chr.11: 72228461-72228461 on Build GRCh38
rs777475102 Chr.11: 72228413-72228413 on Build GRCh38
rs202044589 Chr.11: 72228484-72228484 on Build GRCh38
rs745405566 Chr.11: 72228426-72228426 on Build GRCh38
rs371726889 Chr.11: 72228513-72228513 on Build GRCh38
rs1046707024 Chr.11: 72228416-72228416 on Build GRCh38
rs1000207551 Chr.11: 72228509-72228509 on Build GRCh38
rs966495240 Chr.11: 72228531-72228531 on Build GRCh38
rs916614738 Chr.11: 72228552-72228552 on Build GRCh38
rs779288154 Chr.11: 72228343-72228343 on Build GRCh38
rs759591470 Chr.11: 72228360-72228360 on Build GRCh38
rs145795148 Chr.11: 72228479-72228479 on Build GRCh38
rs887520792 Chr.11: 72228667-72228667 on Build GRCh38
rs760741506 Chr.11: 72228393-72228393 on Build GRCh38
rs769709433 Chr.11: 72228434-72228434 on Build GRCh38
rs371187101 Chr.11: 72228419-72228419 on Build GRCh38
rs949650901 Chr.11: 72228560-72228560 on Build GRCh38
rs765269255 Chr.11: 72228534-72228534 on Build GRCh38
rs531123246 Chr.11: 72228353-72228353 on Build GRCh38
rs755620537 Chr.11: 72228700-72228700 on Build GRCh38
rs549746015 Chr.11: 72228674-72228674 on Build GRCh38
rs779291537 Chr.11: 72228407-72228407 on Build GRCh38
rs994354885 Chr.11: 72228409-72228409 on Build GRCh38
rs748448649 Chr.11: 72228567-72228567 on Build GRCh38
rs1005849096 Chr.11: 72228444-72228444 on Build GRCh38
rs759397146 Chr.11: 72228282-72228282 on Build GRCh38
rs568856228 Chr.11: 72228579-72228579 on Build GRCh38
rs918846796 Chr.11: 72228601-72228601 on Build GRCh38
rs769645440 Chr.11: 72228368-72228368 on Build GRCh38
rs563824788 Chr.11: 72228350-72228350 on Build GRCh38
rs199604909 Chr.11: 72228342-72228342 on Build GRCh38
rs748491872 Chr.11: 72228348-72228348 on Build GRCh38
rs750453091 Chr.11: 72228325-72228325 on Build GRCh38
rs17847221 Chr.11: 72228551-72228551 on Build GRCh38
rs764410685 Chr.11: 72228475-72228475 on Build GRCh38
rs373971775 Chr.11: 72228463-72228463 on Build GRCh38
rs771083237 Chr.11: 72228521-72228521 on Build GRCh38
rs148394356 Chr.11: 72228369-72228369 on Build GRCh38
rs200492197 Chr.11: 72228324-72228324 on Build GRCh38
rs755641145 Chr.11: 72228487-72228487 on Build GRCh38

View Full Product Details