Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95835022-95835520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR2
Entrez Gene ID
8898
Gene Name
myotubularin related protein 2
Gene Aliases
CMT4B, CMT4B1
UniGene
Hs.181326
Gene Chromosome Location
Chr.11: 95832880-95924207 on Build GRCh38
Transcripts
NM_001243571.1 NM_016156.5 NM_201278.2 NM_201281.2 XM_005274374.2 XM_005274375.2 XM_006718934.2 XM_006718935.2 XM_006718936.3 XM_011543058.2 XM_011543059.2 XM_017018517.1 XM_017018518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5730895 c.1671C>A p.A557A Chr.11: 95835335-95835335 on Build GRCh38
1357999 c.1885_1886insAG p.A629fs*>16 Chr.11: 95835336-95835337 on Build GRCh38
1358001 c.1875C>A p.S625S Chr.11: 95835347-95835347 on Build GRCh38
1358000 c.1669_1670insAG p.A557fs*>16 Chr.11: 95835336-95835337 on Build GRCh38
1258039 c.1655C>G p.S552* Chr.11: 95835351-95835351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747684213 Chr.11: 95835338-95835338 on Build GRCh38
rs765751970 Chr.11: 95835403-95835403 on Build GRCh38
rs1045921196 Chr.11: 95835442-95835442 on Build GRCh38
rs578159 Chr.11: 95835205-95835205 on Build GRCh38
rs759874380 Chr.11: 95835213-95835213 on Build GRCh38
rs775643975 Chr.11: 95835364-95835364 on Build GRCh38
rs528877763 Chr.11: 95835215-95835215 on Build GRCh38
rs907160224 Chr.11: 95835173-95835173 on Build GRCh38
rs778784540 Chr.11: 95835465-95835465 on Build GRCh38
rs778356037 Chr.11: 95835437-95835437 on Build GRCh38
rs780878741 Chr.11: 95835337-95835340 on Build GRCh38
rs114078363 Chr.11: 95835127-95835127 on Build GRCh38
rs1004458967 Chr.11: 95835207-95835207 on Build GRCh38
rs761103530 Chr.11: 95835455-95835456 on Build GRCh38
rs766329068 Chr.11: 95835260-95835260 on Build GRCh38
rs753364428 Chr.11: 95835361-95835361 on Build GRCh38
rs745726744 Chr.11: 95835469-95835469 on Build GRCh38
rs748069614 Chr.11: 95835343-95835344 on Build GRCh38
rs3200434 Chr.11: 95835178-95835178 on Build GRCh38
rs986885163 Chr.11: 95835283-95835283 on Build GRCh38
rs762262045 Chr.11: 95835396-95835396 on Build GRCh38
rs574781741 Chr.11: 95835051-95835051 on Build GRCh38
rs570725885 Chr.11: 95835216-95835216 on Build GRCh38
rs374869346 Chr.11: 95835483-95835483 on Build GRCh38
rs760768801 Chr.11: 95835365-95835365 on Build GRCh38
rs772992189 Chr.11: 95835250-95835250 on Build GRCh38
rs770701048 Chr.11: 95835458-95835458 on Build GRCh38
rs886048770 Chr.11: 95835379-95835379 on Build GRCh38
rs776978917 Chr.11: 95835489-95835489 on Build GRCh38
rs532885622 Chr.11: 95835325-95835325 on Build GRCh38
rs911695795 Chr.11: 95835301-95835301 on Build GRCh38
rs762842057 Chr.11: 95835254-95835254 on Build GRCh38
rs954675045 Chr.11: 95835061-95835061 on Build GRCh38
rs377085326 Chr.11: 95835272-95835272 on Build GRCh38
rs1054663282 Chr.11: 95835088-95835088 on Build GRCh38
rs1005974577 Chr.11: 95835196-95835196 on Build GRCh38
rs768921858 Chr.11: 95835488-95835488 on Build GRCh38
rs764652405 Chr.11: 95835456-95835457 on Build GRCh38
rs978145324 Chr.11: 95835062-95835062 on Build GRCh38
rs777498850 Chr.11: 95835340-95835340 on Build GRCh38
rs1047408205 Chr.11: 95835190-95835193 on Build GRCh38
rs369804894 Chr.11: 95835371-95835371 on Build GRCh38
rs923973985 Chr.11: 95835322-95835322 on Build GRCh38
rs544104691 Chr.11: 95835177-95835177 on Build GRCh38
rs769463935 Chr.11: 95835339-95835339 on Build GRCh38
rs958314000 Chr.11: 95835336-95835336 on Build GRCh38
rs760965821 Chr.11: 95835277-95835277 on Build GRCh38
rs762500134 Chr.11: 95835461-95835462 on Build GRCh38
rs367677499 Chr.11: 95835157-95835157 on Build GRCh38
rs749063948 Chr.11: 95835342-95835342 on Build GRCh38
rs1055365050 Chr.11: 95835379-95835379 on Build GRCh38
rs751004861 Chr.11: 95835321-95835321 on Build GRCh38
rs138746296 Chr.11: 95835412-95835412 on Build GRCh38
rs587779387 Chr.11: 95835237-95835237 on Build GRCh38
rs140864250 Chr.11: 95835245-95835245 on Build GRCh38
rs116750638 Chr.11: 95835367-95835367 on Build GRCh38
rs777376551 Chr.11: 95835447-95835447 on Build GRCh38
rs886147288 Chr.11: 95835195-95835195 on Build GRCh38
rs750836112 Chr.11: 95835411-95835411 on Build GRCh38
rs112159344 Chr.11: 95835182-95835182 on Build GRCh38
rs777929624 Chr.11: 95835396-95835408 on Build GRCh38
rs185702937 Chr.11: 95835083-95835083 on Build GRCh38
rs374025895 Chr.11: 95835475-95835475 on Build GRCh38
rs769866540 Chr.11: 95835370-95835371 on Build GRCh38
rs754277194 Chr.11: 95835281-95835281 on Build GRCh38
rs749010549 Chr.11: 95835448-95835448 on Build GRCh38
rs181665205 Chr.11: 95835046-95835046 on Build GRCh38
rs758042172 Chr.11: 95835288-95835289 on Build GRCh38
rs767705774 Chr.11: 95835275-95835275 on Build GRCh38
rs188555540 Chr.11: 95835092-95835092 on Build GRCh38
rs76784113 Chr.11: 95835417-95835417 on Build GRCh38
rs911734753 Chr.11: 95835043-95835043 on Build GRCh38
rs372852182 Chr.11: 95835078-95835078 on Build GRCh38
rs886048769 Chr.11: 95835100-95835100 on Build GRCh38
rs775337882 Chr.11: 95835479-95835479 on Build GRCh38
rs780764646 Chr.11: 95835326-95835326 on Build GRCh38
rs770755992 Chr.11: 95835355-95835355 on Build GRCh38
rs754840148 Chr.11: 95835337-95835370 on Build GRCh38
rs764487267 Chr.11: 95835279-95835279 on Build GRCh38
rs923556491 Chr.11: 95835057-95835057 on Build GRCh38
rs371925152 Chr.11: 95835360-95835360 on Build GRCh38
rs1028344089 Chr.11: 95835256-95835256 on Build GRCh38
rs747889472 Chr.11: 95835244-95835244 on Build GRCh38
rs1030574858 Chr.11: 95835221-95835221 on Build GRCh38
rs974980959 Chr.11: 95835043-95835044 on Build GRCh38
rs576774541 Chr.11: 95835175-95835175 on Build GRCh38
rs564689142 Chr.11: 95835145-95835145 on Build GRCh38
rs751292488 Chr.11: 95835336-95835337 on Build GRCh38
rs749443636 Chr.11: 95835454-95835455 on Build GRCh38
rs774324876 Chr.11: 95835265-95835265 on Build GRCh38
rs779529851 Chr.11: 95835293-95835293 on Build GRCh38
rs754152845 Chr.11: 95835381-95835381 on Build GRCh38
rs779740723 Chr.11: 95835306-95835306 on Build GRCh38
rs757744307 Chr.11: 95835285-95835285 on Build GRCh38
rs755674832 Chr.11: 95835418-95835418 on Build GRCh38

View Full Product Details